Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DBC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537959
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537959 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537959 Supplier Invitrogen™ Supplier No. PA537959
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse, rat, dog, pig and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

DBC1 is located within a chromosomal region that shows loss of heterozygosity in some bladder cancers. It contains a 5' CpG island that may be a frequent target of hypermethylation, and it may undergo hypermethylation-based silencing in some bladder cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DBC1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CCAR2
Gene Accession No. Q8N163, Q8VDP4
Gene Alias 2610301G19Rik; bA574M5.1 (deleted in bladder cancer chromosome region candidate 1 (IB3089A)); BMP/retinoic acid inducible neural specific 1; BMP/retinoic acid-inducible neural-specific protein (BRINP); BMP/retinoic acid-inducible neural-specific protein 1; BMP/retinoic acid-inducible neural-specific protein-1; bone morphogenetic protein/retinoic acid inducible neural-specific 1; bone morphogenic protein/retinoic acid inducible neural specific 1; bone morphogenic protein/retinoic acid inducible neural-specific 1; BRINP; Brinp1; Ccar2; cell cycle activator and apoptosis regulator 2; cell cycle and apoptosis regulator 2; cell cycle and apoptosis regulator protein 2; Cell division cycle and apoptosis regulator protein 2; DBC.1; Dbc1; DBC-1; Dbccr1; DBIRD complex subunit KIAA1967; deleted in bladder cancer 1; deleted in bladder cancer chromosome region candidate 1; deleted in bladder cancer protein 1; Deleted in bladder cancer protein 1 homolog; deleted in breast cancer 1; Deleted in breast cancer gene 1 protein; FAM5A; IB3089A; KIAA1967; mKIAA1967; NET35; p30 DBC; p30 DBC protein; p30DBC; RGD1309922
Gene Symbols CCAR2
Host Species Goat
Immunogen Peptide with sequence C-ESHNRFSATEVTNK, from the internal region of CCAR2 (aa842-855).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219158, 57805
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.