Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DBCO-NHS
GREENER_CHOICESDP

Catalog No. 792125 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-212-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 79-212-5 Supplier Tocris Bioscience™ Supplier No. 7921/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Amine reactive and azide reactive building block

DBCO-NHS is an amine reactive and azide reactive building block. It can be used for copper-free click cycloaddition reactions.

Specifications

Chemical Name or Material 2,5-Dioxo-1-pyrrolidinyl-11,12-didehydro-γ-oxodibenz[b,f]azocine-5(6 H)-butanoate
CAS 1353016-71-3
Quantity 25 mg
Target Buffers, Chelators and Reagents
Molecular Formula C23H18N2O5
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.