Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DBeQ, Tocris Bioscience™

Catalog No. p-4816581
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective and reversible p97 ATPase inhibitor

Selective and reversible inhibitor of p97 ATPase (VCP, IC50 = 1.5 μM). Induces executioner caspases (caspase-3 and caspase-7) rapidly. Blocks the degradation of endoplasmic reticulum-associated degradation (ERAD) reporters; also blocks autophagosome maturation and promotes accumulation of LC3-II.

Chemical Identifiers

CAS 177355-84-9
Molecular Formula C22H20N4
Molecular Weight (g/mol) 340.43
MDL Number MFCD03691820
InChI Key QAIMUUJJAJBPCL-UHFFFAOYSA-N
Synonym dbeq, n2,n4-dibenzylquinazoline-2,4-diamine, n,n'-dibenzylquinazoline-2,4-diamine, 2,4-dibencilaminoquinazoline, 2,4-quinazolinediamine, n,n'-bis phenylmethyl, n*2*,n*4*-dibenzyl-quinazoline-2,4-diamine, n2,n4-bis phenylmethyl-2,4-quinazolinediamine, jrf12, jrf 12, jrf-12
PubChem CID 676352
IUPAC Name N2,N4-dibenzylquinazoline-2,4-diamine
SMILES C(NC1=NC(NCC2=CC=CC=C2)=C2C=CC=CC2=N1)C1=CC=CC=C1

Specifications

Quantity 10 mg
Formula Weight 340.42
Percent Purity >99%
Chemical Name or Material DBeQ
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.