Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DBF4 Antibody (24D8), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP326632100UL
Description
DBF4 Monoclonal antibody specifically detects DBF4 in Human samples. It is validated for Western Blot, ELISA, ImmunocytochemistrySpecifications
DBF4 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunocytochemistry | |
24D8 | |
Western Blot 1:500-1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
ASKchif, DBF4 homolog (S. cerevisiae), DBF4ACHIF, DBF4-type zinc finger-containing protein 1, ZDBF1DBF-type containing 1 | |
A synthesized peptide derived from Human DBF4 [UniProt Q9UBU7] | |
100 μL | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
10926 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction