Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DBF4 Rabbit anti-Human, Clone: S06-6I2, Novus Biologicals™

Rabbit Monoclonal Antibody
$274.50 - $584.50
Specifications
Antigen | DBF4 |
---|---|
Clone | S06-6I2 |
Dilution | Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Monoclonal |
Description
DBF4 Monoclonal antibody specifically detects DBF4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ImmunofluorescenceSpecifications
DBF4 | |
Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ASKchif, DBF4 homolog (S. cerevisiae), DBF4ACHIF, DBF4-type zinc finger-containing protein 1, ZDBF1DBF-type containing 1 | |
A synthetic peptide of human DBF4 (Uniprot#: Q9UBU7) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S06-6I2 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
10926 | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title