Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DBF4A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988105
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8105 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8105 Supplier Bioss Supplier No. BS7895R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen DBF4A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DBF4
Gene Accession No. Q9UBU7
Gene Alias AA545217; Activator of S phase kinase; ASK; CHIF; Chiffon homolog A; Dbf4; DBF4 homolog; DBF4 homolog (S. cerevisiae); DBF4 zinc finger; DBF4 zinc finger A; Dbf4a; DBF4-type zinc finger-containing protein 1; MuDBF4; Protein DBF4 homolog A; RGD1305854; ZDBF1; zinc finger, DBF-type containing 1
Gene Symbols DBF4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DBF4A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10926
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.