Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DBF4B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | DBF4B |
---|---|
Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
DBF4B Polyclonal specifically detects DBF4B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
DBF4B | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
80174 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: HPSQENSFAPADIPVKGPLLFPEARPWLMSARCWVRPFPFVTWGCLIPHDTTPLHEEVSPCPCLRLGYLYLLLTQSLWCRVRVPSLSTAGPIPRTSHPCTLAFPSYLNDHDLGHLCQAKPQGWNTPQPFLHCGFLA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Human | |
Activator of S phase kinase-like protein 1, activator of S-phase kinase-like protein 1, ASKL1CHIFB, ASK-like protein 1, chifb, Chiffon homolog B, DBF4 homolog B (S. cerevisiae), Dbf4-related factor 1, DRF1MGC15009, FLJ13087, protein DBF4 homolog B, ZDBF1B, zinc finger, DBF-type containing 1B | |
DBF4B | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title