Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DBF4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571078
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571078 Supplier Invitrogen™ Supplier No. PA571078
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

This gene encodes a regulator of the CDC7-like 1 protein, a serine-threonine kinase which links cell cycle regulation to genome duplication. This protein localizes to the nucleus and its expression is cell cycle-regulated. Alternative splicing of this gene results in two transcript variants encoding different isoforms. Additional transcript variants have been described, but their full length sequences have not been determined. [provided by RefSeq].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DBF4B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DBF4B
Gene Accession No. Q8NFT6
Gene Alias Activator of S phase kinase-like protein 1; activator of S-phase kinase-like protein 1; ASKL1; ASK-like protein 1; CHIFB; chiffon homolog B; DBF4 homolog B; DBF4 zinc finger B; DBF4B; Dbf4-related factor 1; DRF1; protein DBF4 homolog B; ZDBF1B; zinc finger, DBF-type containing 1B
Gene Symbols DBF4B
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human DBF4B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80174
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.