Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DBF4B Polyclonal Antibody, Abbomax

Catalog No. 502204012
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4012 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4012 Supplier Abbomax Inc Supplier No. 601790
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Spleen Cellular location: Nuclear.

DBF4B Polyclonal antibody specifically detects DBF4B in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen DBF4B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene DBF4B
Gene Accession No. Q8NFT6
Gene Alias Activator of S phase kinase-like protein 1; activator of S-phase kinase-like protein 1; ASKL1; ASK-like protein 1; CHIFB; chiffon homolog B; DBF4 homolog B; DBF4 zinc finger B; DBF4B; Dbf4-related factor 1; DRF1; protein DBF4 homolog B; ZDBF1B; zinc finger, DBF-type containing 1B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the N-terminus of DBF4B protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80174
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.