Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DBI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592426
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592426 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592426 Supplier Invitrogen™ Supplier No. PA592426
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MWGDLWLLPP ASANPGTGTE AEFEKAAEEV RHLKTKPSDE EMLFIYGHYK QATVGDINTE RPGMLDFTGK AKWDAWNELK GTSKEDAMKA YINKVEELKK KYGI; Positive Samples: HT-29, THP-1, MCF7, NCI-H460, A673, U-937, U-251MG; Cellular Location: Endoplasmic reticulum, Golgi apparatus.

This gene encodes diazepam binding inhibitor, a protein that is regulated by hormones and is involved in lipid metabolism and the displacement of beta-carbolines and benzodiazepines, which modulate signal transduction at type A gamma-aminobutyric acid receptors located in brain synapses. The protein is conserved from yeast to mammals, with the most highly conserved domain consisting of seven contiguous residues that constitute the hydrophobic binding site for medium- and long-chain acyl-Coenzyme A esters. Diazepam binding inhibitor is also known to mediate the feedback regulation of pancreatic secretion and the postprandial release of cholecystokinin, in addition to its role as a mediator in corticotropin-dependent adrenal steroidogenesis. Three pseudogenes located on chromosomes 6, 8 and 16 have been identified. Multiple transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DBI
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DBI
Gene Accession No. P07108
Gene Alias ACBD 1; ACBD1; ACBP; Acoabp3; acyl coenzyme A binding protein; acyl-CoA binding protein; acyl-CoA-binding protein; acyl-Coenzyme A binding domain containing 1; CCK RP; CCKRP; CCK-RP; cholecystokinin-releasing peptide, trypsin-sensitive; Dbi; diazepam binding inhibitor; Diazepam binding inhibitor (GABA receptor modulator acyl-Coenxyme A binding protein); diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein); Diazepam binding inhibitor (GABA receptor modulator, acyl-Coenxyme A binding protein); diazepam binding inhibitor (GABA receptor modulator, acyl-Coenzyme A binding protein); diazepam binding inhibitor, acyl-CoA binding protein; diazepam binding inhibitor, splice form 1b; diazepam-binding inhibitor; Endozepine; EP; GABA receptor modulator; LRRGT00046; MGC70414; octadecaneuropeptide; ODN; RNACOABP3; Triakontatetraneuropeptide; Ttn; valium
Gene Symbols DBI
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-104 of human DBI (NP_001171513.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1622
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.