Learn More
3M™ DBI-SALA™ Delta™ Comfort Vest-Style Climbing Harness
Catalog No. 19040601 Shop All 3M Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-040-601 Medium
19-040-617 Small
19-040-618 Large
19-040-619 X-Large
19-040-620 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-040-601 Supplier 3M™ Supplier No. 1100637
Designed to help elevate employees safety and comfort.
  • Features built-in shoulder, back and leg comfort padding
  • Fixed back D-ring
  • Tech-Lite™ quick connect chest and leg straps
  • Durable, lightweight polyester webbing
  • Built-in lanyard keepers with break-away feature
  • Revolver™ vertical torso adjusters
  • Loops for body belt
  • Impact Indicator
  • i-Safe™ equipped with protected labels
  • Recommended industry: Construction, General Industrial, Oil and Gas, Transportation, Utilities
TRUSTED_SUSTAINABILITY
Specifications
Weight (English) 3.65 lb.
D-Ring Location Front
Certifications/Compliance ANSI Z359.1, ANSI Z359.3, OSHA 1910.66, OSHA 1926.502
Size Medium
Webbing Material Polyester