Description
- Features built-in shoulder, back and leg comfort padding
- Fixed back D-ring
- Tech-Lite™ quick connect chest and leg straps
- Durable, lightweight polyester webbing
- Built-in lanyard keepers with break-away feature
- Revolver™ vertical torso adjusters
- Loops for body belt
- Impact Indicator
- i-Safe™ equipped with protected labels
- Recommended industry: Construction, General Industrial, Oil and Gas, Transportation, Utilities
Specifications
Specifications
| Weight (English) | 3.65 lb. |
| D-Ring Location | Front |
| Size | Large |
| Certifications/Compliance | ANSI Z359.1, ANSI Z359.3, OSHA 1910.66, OSHA 1926.502 |
| Webbing Material | Polyester |