Description
- Body belt and side D-rings
- Lightweight polyester webbing
- Plated forged alloy steel hardware
- Unique spring-loaded stand-up dorsal D-ring with impact indicator
- Five-point adjustment
- Nonslip, pass-thru buckle chest strap
Compliance
Meet or exceed ANSI™ Z359 and applicable OSHA regulations.
Specifications
Specifications
| Material | Polyester Webbing |
| Style | Construction Vest |
| D-Ring Location | Back, Side |
| Size | Universal |
| Certifications/Compliance | Meets or exceeds ANSI Z359 and applicable OSHA regulations. |