Description
- No-tangle design
- Stand-up back D-ring
- Tongue or Parachute buckle leg straps
- Repel™ webbing
- Built-in lanyard keepers
- Revolver™ vertical torso adjusters
Specifications
Specifications
| Style | Vest |
| Product Type | Vest-Style Harness |
| Weight (English) | 3.7 lb. |
| Webbing Material | Polyester |
| Size | X-Small |
| D-Ring Location | Back |
| Certifications/Compliance | ANSI A10.32, ANSI Z359.4, OSHA 1926.502, OSHA 1910.66, ANSI Z359.1 |