Description
- No-tangle design
- Stand-up back D-ring
- Tongue or Parachute buckle leg straps
- Repel™ webbing
- Built-in lanyard keepers
- Revolver™ vertical torso adjusters
Specifications
Specifications
| D-Ring Location | Back |
| Size | 2X-Large |
| Certifications/Compliance | Meets or exceeds applicable standards including OSHA/ANSI/ANSIFor Use With (Application) Z359 |
| Product Type | Delta No-Tangle Vest Style Harness w/Back D-ring, Nonslip chest strap w/pass-through buckle |
| Style | Vest |