Learn More
3M™ DBI-SALA™ Delta™ Vest-Style Harness
Catalog No. 19809330 Shop All 3M Products
Change view
Click to view available options
Product Type:
Delta No Tangle Vest Style Harness
Delta No-Tangle Vest Style Harness w/Back D-ring, Nonslip chest strap w/pass-through buckle
Vest-Style Harness
Size:
X-Small
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Size
19-809-330 Vest-Style Harness 2X-Large
19-999-104 Vest-Style Harness 3X-Large
19-809-197 Vest-Style Harness X-Small
19-809-336 Vest-Style Harness X-Large
19-809-238 Delta No-Tangle Vest Style Harness w/Back D-ring, Nonslip chest strap w/pass-through buckle 2X-Large
19-809-211 Delta No Tangle Vest Style Harness X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-809-330 Supplier 3M™ Supplier No. 7100232000
Add to Cart
Add to Cart
Back D-ring, leg straps.
  • No-tangle design
  • Stand-up back D-ring
  • Tongue or Parachute buckle leg straps
  • Repel™ webbing
  • Built-in lanyard keepers
  • Revolver™ vertical torso adjusters
TRUSTED_SUSTAINABILITY
Specifications
Weight (English) 3.7 lb.
Style Vest
Product Type Vest-Style Harness
Size 2X-Large
Certifications/Compliance OSHA 1926.502, OSHA 1910.66
D-Ring Location Back
Webbing Material Repel™ Polyester