Description
- Hybrid shoulder, back and leg comfort padding
- Stand-up Tech-Lite™ aluminum back D-ring
- Duo-Lok™ quick connect chest and leg straps
- Repel™ webbing
- Protective shoulder caps
- Loops for body belt
- Built-in lanyard keepers
- Revolver™ vertical torso adjusters
- Impact Indicator
- Integrated trauma straps
- Recommended Industries: Construction, General Industrial, Oil and Gas, Transportation
Specifications
Specifications
| Certifications/Compliance | ANSI A10.32, ANSI Z359.1, ANSI Z359.4, OSHA 1910.66, OSHA 1926.502 |
| D-Ring Location | Back |
| Weight (English) | 4 lb. |
| Size | Large |