Learn More
3M™ DBI-SALA™ ExoFit NEX™ Vest-Style Harness
Catalog No. 19040614 Shop All 3M Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-040-614 X-Large
19-040-615 2X-Large
19-040-600 Medium
19-040-611 X-Small
19-040-612 Small
19-040-613 Large
19-040-616 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-040-614 Supplier 3M™ Supplier No. 1113010
Advanced vest-style harness designed to help elevate employees safety and comfort.
  • Hybrid shoulder, back and leg comfort padding
  • Stand-up Tech-Lite™ aluminum back D-ring
  • Duo-Lok™ quick connect chest and leg straps
  • Repel™ webbing
  • Protective shoulder caps
  • Loops for body belt
  • Built-in lanyard keepers
  • Revolver™ vertical torso adjusters
  • Impact Indicator
  • Integrated trauma straps
  • Recommended Industries: Construction, General Industrial, Oil and Gas, Transportation
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI A10.32, ANSI Z359.1, ANSI Z359.4, OSHA 1910.66, OSHA 1926.502
Size X-Large
Weight (English) 4 lb.
D-Ring Location Back