Learn More
3M™ DBI-SALA™ ExoFit™ X300 Comfort Construction Climbing/Positioning Safety Harness
The 3M™ DBI-SALA™ ExoFit™ X300 is designed for those who refuse to compromise. The X300 harnesses are outfitted with high-quality components that have been developed to enhance the safety, convenience and performance for working at height on your job site.
$816.97 - $850.40
Specifications
Buckle Style (Chest) Auto-Locking Quick-Connect
Color Gray
Material Polyester
Product Type Climbing/Positioning Safety Harness
D-Ring Location Back, Front, Hip
View More Specs
Alert:
For industrial/occupational use only. Not for consumer sale or use.
Products 10
Catalog Number Mfr. No. Size Buckle Style (Leg) Length (Metric) Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Size Buckle Style (Leg) Length (Metric) Length (English) Price Quantity & Availability  
13-051-021
View Documents
3M™
7100157134
X-Small Auto-Locking Quick-Connect 46.7614 cm 18.41 in.
Each for $850.40
Only null left
Add to Cart
13-051-121
View Documents
3M™
7012816548
X-Small Tongue Buckle 48.26 cm 19.0 in.
Each for $816.97
Only null left
Add to Cart
13-051-069
View Documents
3M™
7100188762
Small Auto-Locking Quick-Connect 48.26 cm 19.0 in.
Each for $850.40
Only null left
Add to Cart
13-051-122
View Documents
3M™
7012816550
Small Tongue Buckle 46.99 cm 18.5 in.
Each for $816.97
Only null left
Add to Cart
13-051-070
View Documents
3M™
7100157135
Medium Auto-Locking Quick-Connect 45.72 cm 18.0 in.
Each for $850.40
Only null left
Add to Cart
13-051-123
View Documents
3M™
7012816552
Medium Tongue Buckle 48.26 cm 19.0 in.
Each for $816.97
Only null left
Add to Cart
13-051-124
View Documents
3M™
7012816554
Large Tongue Buckle 48.26 cm 19.0 in.
Each for $816.97
Only null left
Add to Cart
13-051-125
View Documents
3M™
7012816556
X-Large Tongue Buckle 48.26 cm 19.0 in.
Each for $816.97
Only null left
Add to Cart
13-051-073
missing translation for 'viewDocuments'
3M™
7012816226
2X-Large Auto-Locking Quick-Connect 44.45 cm 17.5 in.
Each for $850.40
Only null left
Add to Cart
13-051-126
missing translation for 'viewDocuments'
3M™
7012816558
2X-Large Tongue Buckle 48.26 cm 19.0 in.
Each for $816.97
Only null left
Add to Cart
Description
Description
  • The X300 comfort harness is built with 6000 lb. polyester webbing, robust nylon/polyester hybrid comfort padding with reflective 3M Scotchlite™ panels, and lightweight aluminum hardware
  • Revolving Torso Adapters adjust harness fit with ease and speed and lock webbing into place to maintain a comfortable and safe harness fit
  • Equipped with anti-sliding dorsal D-ring pad to help greatly reduce dorsal D-ring slide
  • Integrated easy-link SRL adapter lets you quickly and easily connect and disconnect personal SRLs
  • Auto-locking quick connect buckles secure contact with webbing, reducing the need to readjust
  • Automatic stand-up D-ring (lightweight aluminum) enables efficient dorsal connections
  • Low-profile suspension trauma strap design integrated between harness webbing
  • Equipped with auto-resetting lanyard keepers for fast and easy parking of snap hooks and carabiners
  • 3M™ Connected Safety ID (RFID) integrated for simplified inventory, inspection, and record management
Specifications
Specifications
Auto-Locking Quick-Connect
Polyester
Back, Front, Hip
Gray
Climbing/Positioning Safety Harness
Videos
SDS
Documents
Documents
Product Certifications
3M industrial and occupational products are intended, labeled, and packaged for sale to trained industrial and occupational customers for workplace use. Unless specifically stated otherwise on the applicable product packaging or literature, these products are not intended, labeled, or packaged for sale to or use by consumers (e.g., for home, personal, primary or secondary school, recreational/sporting, or other uses not described in the applicable product packaging or literature), and must be selected and used in compliance with applicable health and safety regulations and standards (e.g., U.S. OSHA, ANSI), as well as all product literature, user instructions, warnings, and other limitations, and the user must take any action required under any recall, field action, or other product use notice. Misuse of 3M industrial and occupational products may result in injury, sickness, death, or property damage. For help with product selection and use, consult your on-site safety professional, industrial hygienist, or other subject matter expert.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.