Learn More
3M™ DBI-Sala™ Hi-Vis WorkVest and Harness Combination
Catalog No. 19047702 Shop All 3M Products
Change view
Click to view available options
Size:
Large Vest; Universal Harness
Medium Vest; Universal Harness
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-047-702 Medium Vest; Universal Harness
19-059-321 Large Vest; Universal Harness
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-047-702 Supplier 3M™ Supplier No. 7012815583
Add to Cart
Add to Cart
Combination work vest and fall protection harness
Combines high-visibilty work vest with a full-body harness that safely distributes fall arrest forces throughout the body.
  • Vest is made of certified hi-vis fabric with reflective tape
  • Delta No-Tangle™ harness design holds its shape, makes donning easy
  • Highly durable polyester webbing has corrosion-resistant coated hardware
  • Includes padded shoulders, neck liner, belt loops for a body belt, and convenient, Velcro™ tool fasteners for attaching accessories
  • With two chest pockets, one with slots for pens and pencils, one with a snap closure to prevent loss of expensive items
Compliance
Meets or exceeds OSHA, ANSI Z359.1, and ANSI/ISEA 107–1999 standards.
TRUSTED_SUSTAINABILITY
Specifications
Color Hi-vis Orange
Size Medium Vest; Universal Harness
Certifications/Compliance Meets or exceeds OSHA, ANSI Z359.1, and ANSIFor Use With (Application)/ISEA 107-1999 standards.
D-Ring Location Back
Buckle Style Tongue