Learn More
Description
- PVC davit mast sleeve liner
- Set-up and dismantle without tools
- Lightweight, corrosion resistant aluminum construction
Specifications
Specifications
| For Use With (Equipment) | Fits 22 to 24 in. (55.8 to 60.9 cm) diameter opening |
| Certifications/Compliance | OSHA 1910.66 |
| Weight (English) | 37 lb. |
| Product Type | Confined Space Manhole Collar Davit Base |
| Weight (Metric) | 16.8 kg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.