Learn More
3M™ DBI-SALA™ Delta™ Tower Climbing Harness
Catalog No. 19809232 Shop All 3M Products
Change view
Click to view available options
D-Ring Location:
Back, Front, Side, Suspension
Back, Side, Front, Suspension
Size:
Small
Medium
Large
X-Large
3X-Large
Style:
Cross-over
Vest
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. D-Ring Location Size Style
19-809-232 Back, Front, Side, Suspension Medium Cross-over
19-809-250 Back, Front, Side, Suspension Large Vest
19-809-251 Back, Front, Side, Suspension Medium Vest
19-809-252 Back, Front, Side, Suspension X-Large Vest
19-809-228 Back, Side, Front, Suspension 3X-Large Cross-over
19-809-230 Back, Front, Side, Suspension Small Cross-over
19-809-227 Back, Front, Side, Suspension X-Large Cross-over
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-809-232 Supplier 3M™ Supplier No. 1103351
Back, front and side D-rings, belt with pad, seat sling with positioning D-rings, tongue buckle leg straps.
  • No-tangle design
  • Stand-up back D-ring
  • Front D-ring
  • Body belt/hip pad with side D-rings
  • Tool Pouch D-rings
  • Seat sling with positioning D-rings
TRUSTED_SUSTAINABILITY
Specifications
Style Cross-over
Size Medium
Webbing Material Polyester
D-Ring Location Back, Front, Side, Suspension
Weight (English) 7.6 lb.
Product Type Cross-Over Style Tower Climbing Harness