Learn More
Description
- Detent style attachment
- Set-up and dismantle without tools
- Durable and corrosion resistant construction
Specifications
Specifications
| Length (Metric) | 1.2 m |
| Weight (English) | 27.8 lb. |
| Weight (Metric) | 12.6 kg |
| Product Type | Confined Space Vehicle Hitch Mount Sleeve Extension |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.