Learn More
3M™ DBI-SALA™ Vest-Type Harnesses
Catalog No. 18704 Shop All 3M Products
Change view
Click to view available options
Buckle Style:
Parachute, Tech-Lite™ Quick Connect, Pass-Thru
Tongue buckle
D-Ring Location:
Back
Back, Side
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Buckle Style D-Ring Location
18-704 Tongue buckle Back
19-070-700 Parachute, Tech-Lite™ Quick Connect, Pass-Thru Back
19-070-748 Tongue buckle Back, Side
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-704 Supplier 3M™ Supplier No. 7100232005
Add to Cart
Add to Cart
The culmination of more than twenty years of design, research, manufacturing know-how and on-the-job performance. Featuring coated, corrosion-resistant hardware, water-repellant and abrasion-resistant webbing and a build quality that is second to none.
  • No-tangle design
  • Stand-up back D-ring
  • Repel™ webbing
  • Built-in lanyard keepers
  • Revolver™ vertical torso adjusters
  • Impact Indicator
Order Info
One size fits most workers. Choice of leg strap buckles.
Compliance
Meet ANSI™ Z359.1-1992, ANSI A10.14-1991, and OSHA requirements.
TRUSTED_SUSTAINABILITY
Specifications
Material Polyester
Webbing Material Repel™ Polyester
D-Ring Location Back
Weight (English) 3.7 lb.
Capacity (English) 420 lb.
Capacity (Metric) 190 kg
For Use With (Application) Suitable for fall arrest or restraint.
Style Vest
Certifications/Compliance Meets ANSI Z359.1-1992, ANSIFor Use With (Application) A10.14-1991, OSHA
Size Universal
Buckle Style Tongue buckle