Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DBNDD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560575
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560575 Supplier Invitrogen™ Supplier No. PA560575
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VNVDTLEQVE LIDLGDPDAA DVFLPCEDPP PTPQSSGVDN HLEELSLPVP TSDRTTSRTS S Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 85%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen DBNDD2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DBNDD2
Gene Accession No. Q9BQY9
Gene Alias 1110017A21Rik; 2900022J10Rik; AU041050; AW048677; C20orf35; casein kinase-1 binding protein; CK1BP; D2Bwg0891e; Dbndd2; dysbindin (dystrobrevin binding protein 1) domain containing 2; dysbindin domain containing 2; dysbindin domain-containing protein 2; HSMNP1; likely orthologue of H. sapiens chromosome 20 open reading frame 35; Nkip; R74724; SCF apoptosis response protein 1
Gene Symbols DBNDD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DBNDD2. Recombinant protein control fragment (Product #RP-99473).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55861
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.