Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540501
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540501 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540501 Supplier Invitrogen™ Supplier No. PA540501
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PKEPASCLLK EKERKAALPA ATTPGPGLET AGPADAPAGA VVGGGSPRGR Sequence homology: Cow: 85%; Dog: 85%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 83%; Rat: 83%; Sheep: 85%.

GC (vitamin D-binding protein) is involved in vitamin D transport/storage, scavenging of extracellular G-actin, enhancement of the chemotactic activity of C5 alpha for neutrophils in inflammation and macrophage activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DBP
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GC
Gene Accession No. P02774
Gene Alias DBP; DBP/ VDBG; DBP/GC; DBP02; DBP-maf; epididymis secretory protein Li 51; Gc; Gc globulin; Gc protein-derived macrophage activating factor; GC, vitamin D binding protein; gc-globulin; GcMAF; Gc-MAF; GRD3; group specific component; Group specific component vitamin D binding protein; Group-specific component; group-specific component (vitamin D binding protein); hDBP; HEL-S-51; V VDB; VDB; VDBG; Vdbp; Vitamin D binding alpha globulin; vitamin D-binding alpha-globulin; vitamin D-binding protein; Vitamin D-binding protein-macrophage activating factor
Gene Symbols GC
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human DBP (aa 41-90).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2638
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.