Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DBPA Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171827AF647
Description
DBPA Polyclonal antibody specifically detects DBPA in Human samples. It is validated for Western BlotSpecifications
DBPA | |
Polyclonal | |
Western Blot | |
cold shock domain protein A, Cold shock domain-containing protein A, cold-shock domain protein A, CSDA1, dbpA, DBPAcold-shock domain containing A1, DNA-binding protein A, Single-strand DNA-binding protein NF-GMB, ZO-1-associated nucleic acid-binding protein, ZONAB | |
The immunogen for this product maps to a region between residue 250 and 300 of human ZO-1-associated nucleic acid-binding protein using the numbering given in entry NP_003642.3 (GeneID 8531). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
8531 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction