Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DBR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530616
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530616 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530616 Supplier Invitrogen™ Supplier No. PA530616
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549. Predicted reactivity: Mouse (90%), Rat (90%), Chicken (81%), Rhesus Monkey (100%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The RNA lariat debranching enzyme, or DBR1, specifically hydrolyzes 2-prime-to-5-prime branched phosphodiester bonds at the branch point of excised lariat intron RNA and converts them into linear molecules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DBR1
Applications Western Blot
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene DBR1
Gene Accession No. Q9UK59
Gene Alias AW018415; Dbr1; debranching enzyme homolog 1; debranching enzyme homolog 1 (S. cerevisiae); debranching RNA lariats 1; lariat debranching enzyme; RNA lariat debranching enzyme
Gene Symbols DBR1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DBR1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51163
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.