Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DBX1 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281711FR
This item is not returnable.
View return policy
Description
DBX1 Polyclonal antibody specifically detects DBX1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
DBX1 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
120237 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 680 | |
developing brain homeobox 1, developing brain homeobox protein 1, homeobox protein DBX1 | |
DBX1 antibody was raised against a 15 amino acid synthetic peptide near the center of human DBX1. The immunogen is located within amino acids 100 - 150 of DBX1. Amino Acid Squence: AAILSSGPRTETSPAL | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction