Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DBX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572697
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572697 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572697 Supplier Invitrogen™ Supplier No. PA572697
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DBX1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Could have a role in patterning the central nervous system during embryogenesis. Has a key role in regulating the distinct phenotypic features that distinguish two major classes of ventral interneurons, V0 and V1 neurons. Regulates the transcription factor profile, neurotransmitter phenotype, intraspinal migratory path and axonal trajectory of V0 neurons, features that differentiate them from an adjacent set of V1 neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DBX1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene DBX1
Gene Accession No. A6NMT0, P52950, Q5NSW5
Gene Alias AI426026; Dbx; Dbx1; developing brain homeobox 1; developing brain homeobox protein 1; Homeobox protein DBX1; Mmox C
Gene Symbols DBX1
Host Species Rabbit
Immunogen DBX1 antibody for a 15 amino acid synthetic peptide near the center of human DBX1. The immunogen is within amino acids 100 - 150 of DBX1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120237, 13172, 292934
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.