Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DBX2 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282083UV
This item is not returnable.
View return policy
Description
DBX2 Polyclonal antibody specifically detects DBX2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| DBX2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 440097 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| developing brain homeobox 2, developing brain homeobox protein 2, FLJ16139, homeobox protein DBX2 | |
| DBX2 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human DBX2. The immunogen is located within amino acids 30 - 80 of DBX2. Amino Acid Squence: APTPRLQPPAPHDPAT | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction