Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DBX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564261
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564261 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564261 Supplier Invitrogen™ Supplier No. PA564261
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSPRWRENSP EPSERLIQES SGAPPPEANS LQGALYLCSE EEAGSKGVLT G Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 53%.

DBX2 is a member of the developing brain homeobox (DBX) protein family, but while the related protein DBX1 is expressed in various regions of the developing brain, DBX2 shows a more restricted pattern of expression in the brain, and is also expressed in some mesenchymal cells such as limb buds and tooth germs. It is thought that DBX1 and DBX2 promote the development of a subset of interneurons, some of which help mediate left-right coordination of locomotor activity. In Xenopus, DBX2 is involved in primary neurogenesis and early neural plate patterning, and is thought to act as a cross-repressive partner of NKX6-2 in the patterning of the ventral neural tube.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DBX2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DBX2
Gene Accession No. Q6ZNG2
Gene Alias 9430056A22Rik; 9630005K15; AI846217; DBX2; developing brain homeobox 2; developing brain homeobox protein 2; Gm1229; Homeobox protein DBX2
Gene Symbols DBX2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DBX2. Recombinant protein control fragment (Product #RP-103622).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440097
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.