Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DBZ
GSA_VAGREENER_CHOICE

Catalog No. 448910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-891-0 10 mg
44-895-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-891-0 Supplier Tocris Bioscience™ Supplier No. 4489/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

γ-secretase inhibitor; inhibits Notch pathway

DBZ is an inhibitor of γ-secretase aspartyl protease. DBZ blocks amyloid precursor protein-like (APPL) and Notch cleavage (IC50 values are 2.6 nM and 1.7-2.9 nM, respectively). By inhibiting Notch and the Notch pathway, DBZ improves efficiency of iPSC generation and production of iPSCs from mouse and human keratinocytes without KLF4 and CMYC. In an animal model of Alzheimer's disease, DBZ inhibits the cleavage of amyloid precursor protein into Aβ40. DBZ promotes cochlear supporting cell proliferation and hair cell mitotic regeneration in neonatal mice. DBZ in combination with iDOT1L enables maintenance of human na?ve-state pluripotency in non-hypoxic conditions.

DBZ synthesized to Ancillary Material Grade also available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Chemical Name or Material DBZ
CAS 209984-56-5
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C26H23F2N3O3
Formula Weight 463.48
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.