Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DC 260126
GREENER_CHOICESDP

Catalog No. 535750 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-575-0 50 mg
53-571-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-575-0 Supplier Tocris Bioscience™ Supplier No. 5357/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

FFA1 (GPR40) antagonist

DC 260126 is a free fatty acid receptor 1 (FFA1/GPR40) antagonist. Inhibits FFA-induced increases in intracellular Ca2+ levels and suppresses palmitic acid potentiated glucose-stimulated insulin secretion in Min6 pancreatic β cells in vitro. Decreases serum insulin levels, improves insulin sensitivity and reduces the number of apoptotic pancreatic β cells in obese diabetic (db/db) rats.

Specifications

Chemical Name or Material N-(4-Butylphenyl)-4-fluorobenzenesulfonamide
CAS 346692-04-4
Quantity 50 mg
Target Free Fatty Acid Receptor Antagonists
Molecular Formula C16H18FNO2S
Formula Weight 307.38
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.