Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DC-LAMP Antibody (007), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289553AF647
Description
DC-LAMP Monoclonal specifically detects DC-LAMP in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
DC-LAMP | |
Monoclonal | |
Alexa Fluor 647 | |
CD208 antigen, DC-LAMP, DCLAMPDC-lysosome-associated membrane glycoprotein, LAMP, LAMP-3, lysosomal-associated membrane protein 3DC LAMP, lysosome-associated membrane glycoprotein 3, Protein TSC403, TSC403CD208 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human DC-LAMP (NP_055213.2; Lys28-Thr381). | |
0.1 mL | |
Dendritic Cell Markers | |
27074 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
7 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction