Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DC-LAMP Antibody (104G4), Alexa Fluor™ 546, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals DDX0190A546100
Description
DC-LAMP Monoclonal specifically detects DC-LAMP in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
DC-LAMP | |
Monoclonal | |
0.5mg/mL | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
CD208 antigen, DC-LAMP, DCLAMPDC-lysosome-associated membrane glycoprotein, LAMP, LAMP-3, lysosomal-associated membrane protein 3DC LAMP, lysosome-associated membrane glycoprotein 3, Protein TSC403, TSC403CD208 | |
Mouse | |
Ion exchange chromatography | |
RUO | |
Primary | |
Human, Sheep | |
IgG1 |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
104G4 | |
Alexa Fluor 546 | |
NP_005502 | |
LAMP3 | |
DC-LAMP/CD208 was used as immunogen. | |
0.1 mg | |
Dendritic Cell Markers | |
27074 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction