Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DC-LAMP Antibody (L3P2H10*E3), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250464AF532
Description
DC-LAMP Monoclonal specifically detects DC-LAMP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
DC-LAMP | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
LAMP3 | |
Ovalbumin-conjugated synthetic peptide - RCQSSGYQRI | |
0.1 mL | |
Primary | |
Human | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
L3P2H10*E3 | |
Western Blot, ELISA, Immunohistochemistry | |
CD208 antigen, DC-LAMP, DCLAMPDC-lysosome-associated membrane glycoprotein, LAMP, LAMP-3, lysosomal-associated membrane protein 3DC LAMP, lysosome-associated membrane glycoprotein 3, Protein TSC403, TSC403CD208 | |
Mouse | |
Protein A purified | |
Dendritic Cell Markers | |
27074.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction