Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DC-SIGN/CD209 Antibody (061), DyLight 550

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289332R
Description
DC-SIGN/CD209 Monoclonal specifically detects DC-SIGN/CD209 in Human samples. It is validated for Western Blot, ELISA.Specifications
DC-SIGN/CD209 | |
Monoclonal | |
DyLight 550 | |
CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human DC-SIGN/CD209 (NP_066978.1; Lys62-Ala404). | |
0.1 mL | |
Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
30835 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
61 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction