Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DC-SIGN/CD209 Antibody (109H12.03), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals DDX0207A647
Description
DC-SIGN/CD209 Monoclonal specifically detects DC-SIGN/CD209 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface).Specifications
DC-SIGN/CD209 | |
Monoclonal | |
Alexa Fluor 647 | |
NP_066978 | |
CD209 | |
human DC-SIGN-transfected HeLa cells | |
0.1 mg | |
Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
30835 | |
Store at -20C in the dark. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunohistochemistry | |
109H12.03 | |
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500 | |
CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 | |
Mouse | |
Ion exchange chromatography | |
RUO | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction