Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DC-SIGN/CD209 Antibody (120507) [Biotin], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB161B
Description
DC-SIGN/CD209 Monoclonal antibody specifically detects DC-SIGN/CD209 in Human samples. It is validated for Western Blot, Flow Cytometry, Adhesion Blockade, CyTOFSpecifications
| DC-SIGN/CD209 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF | |
| 120507 | |
| Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF-ready | |
| CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 | |
| NIH-3T3 mouse embryonic fibroblast cell line transfected with human DC-SIGN/CD209 | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
| 30835 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction