Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DC-SIGN/CD209 Antibody (120507) [Janelia Fluor™ 646], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB161J
Description
DC-SIGN/CD209 Monoclonal antibody specifically detects DC-SIGN/CD209 in Human samples. It is validated for Western Blot, Flow Cytometry, Adhesion Blockade, CyTOFSpecifications
DC-SIGN/CD209 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF | |
120507 | |
Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF-ready | |
CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 | |
NIH-3T3 mouse embryonic fibroblast cell line transfected with human DC-SIGN/CD209 | |
0.1 mL | |
Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
30835 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction