Learn More
DC-SIGN/CD209 Antibody (LWC06), Alexa Fluor™ 488, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP143328AF488
Catalog No. NBP143328AG
Only null left
Add to Cart
Description
Description
DC-SIGN/CD209 Monoclonal specifically detects DC-SIGN/CD209 in Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
Specifications
DC-SIGN/CD209
Monoclonal
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
CD209
The immunogen for this antibody was CIRE.
0.1 mL
Adaptive Immunity, Immunology, Virology Bacteria and Parasites
30835
Store at 4C in the dark.
IgG2a
Western Blot, Flow Cytometry
LWC06
Western Blot, Flow Cytometry
CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1
Rat
Protein A or G purified
RUO
Primary
Mouse
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.