Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DC-SIGN/CD209 Antibody (MM0239-4G10) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
Antigen | DC-SIGN/CD209 |
---|---|
Clone | MM0239-4G10 |
Concentration | LYOPH |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
DC-SIGN/CD209 Monoclonal specifically detects DC-SIGN/CD209 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
DC-SIGN/CD209 | |
LYOPH | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
Q9NNX6 | |
30835 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
MM0239-4G10 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 | |
CD209 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title