Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DC-SIGN+DC-SIGNR Antibody (DC28), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB16211P
Description
DC-SIGN+DC-SIGNR Monoclonal antibody specifically detects DC-SIGN+DC-SIGNR in Human samples. It is validated for Western Blot, Flow Cytometry, Adhesion Blockade, CyTOFSpecifications
DC-SIGN+DC-SIGNR | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
30835 | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF | |
DC28 | |
Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF-ready | |
CD209,CD209 antigen,CDSIGN,CLEC4L,DC-SIGN,DC-SIGN1,hDC-SIGN | |
E. coli-derived recombinant human DC-SIGN, Extracellular domain | |
0.5 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction