Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DC-SIGNR/CD299/CLEC4M Antibody (MM0241-2U25) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. p-7199592 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019075 0.025 mg
NBP212158 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019075 Supplier Novus Biologicals Supplier No. NBP2121580.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DC-SIGNR/CD299/CLEC4M Monoclonal specifically detects DC-SIGNR/CD299/CLEC4M in Human samples. It is validated for Flow Cytometry, CyTOF-ready, Block/Neutralize.

Specifications

Antigen DC-SIGNR/CD299/CLEC4M
Applications Blocking Assay, Flow Cytometry, CyTOF
Classification Monoclonal
Clone MM0241-2U25
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000, Blocking/Neutralizing 50-200 molar excess, CyTOF-ready
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. NP_001138382
Gene Alias CD209L1, CD209Ldendritic cell-specific ICAM-3-grabbing nonintegrin 2, CD299, CD299 antigenMGC129964, C-type lectin domain family 4, member M, DC-SIGN2CD209 antigen-like protein 1, DC-SIGNRDC-SIGN-related protein, DCSIGNRMGC47866, Dendritic cell-specific ICAM-3-grabbing non-integrin 2, HP10347, liver/lymph node-specific ICAM-3 grabbing non-integrin, Liver/lymph node-specific ICAM-3-grabbing non-integrin, L-SIGN, LSIGNC-type lectin domain family 4 member M, mannose binding C-type lectin DC-SIGNR
Gene Symbols CLEC4M
Host Species Mouse
Immunogen Human CLEC4M
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10332
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.