Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DC-STAMP/TM7SF4 Antibody (788524), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB7824F
Description
DC-STAMP/TM7SF4 Monoclonal antibody specifically detects DC-STAMP/TM7SF4 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
DC-STAMP/TM7SF4 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
81501 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, CyTOF | |
788524 | |
Flow Cytometry, CyTOF-ready | |
dendritic cell-specific transmembrane protein, transmembrane 7 superfamily member 4 | |
E. coli-derived recombinant human DC-STAMP, Asp314-Thr376, Accession # Q9H295 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction