Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DC8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561088
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561088 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561088 Supplier Invitrogen™ Supplier No. PA561088
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FVQKLGDALP EEIREPALRD AQWTFESAVQ ENISINGQAW QEASDNCFMD SDIKVLEDQF DEIIVDIATK RKQYP Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 79%.

Part of the MIS12 complex which is required for normal chromosome alignment and segregation and kinetochore formation during mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DC8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NSL1
Gene Accession No. Q96IY1
Gene Alias 4833432M17Rik; AI451597; C1orf48; DC31; DC8; Gm105; Gm984; kinetochore-associated protein NSL1 homolog; MIS14; NSL1; NSL1, MIND kinetochore complex component, homolog; NSL1, MIND kinetochore complex component, homolog (S. cerevisiae); NSL1, MIS12 kinetochore complex component; RGD1562052
Gene Symbols NSL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DC8. Recombinant protein control fragment (Product #RP-94129).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25936
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.