Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DC8 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501558463
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
50-155-8463 100 μL
1 options

Catalog No. 50-155-8463

Supplier: Bethyl Laboratories, Inc A300795A

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Part of the MIS12 complex which is required for normal chromosome alignment and segregation and kinetochore formation during mitosis.

Specifications

Antigen DC8
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene NSL1
Gene Accession No. Q96IY1
Gene Alias DC8, C1orf48, MIS14, kinetochore-associated protein NSL1 homolog
Gene Symbols NSL1
Host Species Rabbit
Immunogen Between 230 and C-term
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25936
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.