Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCAF11 Antibody, Novus Biologicals™
SDP

Catalog No. NB034871 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB034871 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB034871 Supplier Novus Biologicals Supplier No. NBP287249
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DCAF11 Polyclonal specifically detects DCAF11 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen DCAF11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias DDB1 and CUL4 associated factor 11, DDB1- and CUL4-associated factor 11, GL014, PRO2389, WD repeat domain 23, WD repeat-containing protein 23, WDR23DKFZp779A1629
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of DCAF11
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80344
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.