Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCAF12 Antibody, PerCP, Novus Biologicals™

Chicken Polyclonal Antibody
Supplier: Novus Biologicals NBP177373PCP
Description
DCAF12 Polyclonal antibody specifically detects DCAF12 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
DCAF12 | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
DDB1- and CUL4-associated factor 12, CT102, DDB1 and CUL4 associated factor 12, KIAA1892, TCC52, WDR40A | |
Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human TCC52. The immunogen is located within amino acids 390 - 440 of TCC52. Amino Acid Squence: LNHDETWRNYFSDID | |
0.1 mL | |
Cell Biology | |
25853 | |
Store at 4C in the dark. | |
IgY |
Western Blot, ELISA, Immunocytochemistry | |
PerCP | |
PBS | |
Chicken | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction